Find methods for your needs
AN342: Determination of PCBs in Large-Volume Fish Tissue Samples Using Accelerated Solvent Extraction (ASE)
# | Compound Name | Compound Class |
---|---|---|
1 | 2,2',3,3',4,5',6,6-Octachlorobiphenyl | PCB |
2 | 2,2',3,4',5,6,6'-Heptachlorobiphenyl | PCB |
3 | 2,2',3,4,5'-Pentachlorobiphenyl | PCB |
4 | 2,2',4,4',5,6'-Hexachlorobiphenyl | PCB |
5 | 2,2',4,6-Tetrachlorobiphenyl | PCB |
6 | 2,3-Dichlorobiphenyl | PCB |
7 | 2,4,5-Trichlorobiphenyl | PCB |
8 | 2-Chlorobiphenyl | PCB |
9 | Decachlorobiphenyl | PCB |
Description
PCBs are a persistent organic pollutant (POP) and therefore are monitored in marine life including fish. Automated Solvent Extraction (ASE 350) technology, an automated extraction method, has been shown to produce good recoveries of naturally occurring PCBs from fish tissue samples, and is approved for use in U.S. EPA SW-846 Method 3545 for the extraction of PCBs, OCPs, BNAs, OPPs, herbicides, and dioxins and furans. ASE operates at elevated temperatures, thus increasing the efficiency of the extraction process. Here we demonstrate the extraction of PCBs in a raw fish tissue sample.Market: | Food and Beverage |
Keywords: | PCB, Polychlorinated biphenyls (PCBs), Accelerated solvent extraction, ASE, Polychlorinated biphenyls, ASE 200, ASE 350, EPA Method 3545, PCBs, fish tissue, POPs, Decachlorobiphenyl, Octachlorobiphenyl, Heptachlorobiphenyl, Hexachlorobiphenyl, Pentachlorobiphenyl, Tetrachlorobiphenyl, Trichlorobiphenyl, Dichlorobiphenyl, Chlorobiphenyl |
Matrix: | Cod fillet, Raw fish tissue |
For Research Use Only. Not for use in diagnostic procedures.