Find methods for your needs
Separation of related impurities of Calcitonin (Polypeptide Hormone).
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the separation of related impurities of Calcitonin. The separation was performed on a Thermo Scientific Acclaim 120-C18 HPLC column with UV detection at 220 nm. With a total run time of 75 minutes the method allows separation of all related impurities of Calcitonin.| Market: | Pharma |
| Keywords: | Acclaim 120 C18 Column, HPLC, UV detection, Calcitonin, Thyrocalcitonin, Polypeptide hormone |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.