Find methods for your needs
Fast analysis of benzodiazepines using a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of seven benzodiazepines. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than seven minutes, with UV detection at 235 nm.
Market: Pharma
Keywords: benzodiazepines, Diazepam, HPLC, Hypersil GOLD, Lorazepam, Medazepam, Nitrazepam, Nordiazepam, Oxazepam, Temazepam, UV detection
Matrix: N/A
Uploaded on 8/23/2012.
For Research Use Only. Not for use in diagnostic procedures.