Find methods for your needs
Fast analysis of benzodiazepines using a Thermo Scientific Hypersil GOLD HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of seven benzodiazepines. The separation is performed on a Thermo Scientific Hypersil GOLD HPLC column in less than seven minutes, with UV detection at 235 nm.| Market: | Pharma |
| Keywords: | benzodiazepines, Diazepam, HPLC, Hypersil GOLD, Lorazepam, Medazepam, Nitrazepam, Nordiazepam, Oxazepam, Temazepam, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.