Find methods for your needs
AN329: Determination of Total Fat in Powdered Infant Formula Using Accelerated Solvent Extraction (ASE)
Description
The U.S. Nutritional Labeling and Education Act of 1990 requires that food products carry labels listing the content of saturated and unsaturated fats. Accurate determination of fat in certain foods is difficult due to the binding or entrapment of the fat by the matrix. Accelerated Solvent Extraction (ASE) is a new automated extraction technique that can significantly reduce extraction time and solvent consumption and yields equivalent results, without requiring a pretreatment step. In this application note, fat is extracted from infant formulas by ASE without aggressive pretreatments.| Market: | Food and Beverage |
| Keywords: | FAME, Fat, Accelerated solvent extraction, ASE, Fatty acid methyl esters, Infant Formula, Fatty Acid Methyl Esters (FAMEs), ASE 200, ASE 350, Total Fat, AOAC 933.05, AOAC 905.02, AOAC 989.05 |
| Matrix: | Powdered infant formula |
For Research Use Only. Not for use in diagnostic procedures.