Find methods for your needs
Separation of related Impurities of Ondansetron
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied to the separation of the related impurities of Ondansetron. The separation was performed on a Thermo Scientific Acclaim Polar Advantage II C18 HPLC column with UV detection at 216 nm. With a total run time of 65 minutes the method gives separation of all related impurities of Ondansetron.| Market: | Pharma |
| Keywords: | HPLC, Ondansetron, UV detection, Impurity profile, Acclaim Polar Advantage-II, Imidazole, 2-Methyl Imidazole, Desmethyl Ondansetron, 4-Oxo Carbazole |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.