Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied to the estimation of Dicyandiamide. The analysis was performed on a Thermo Scientific Hypersil Gold aQ HPLC column with UV detection at 215 nm. With a total run time of 10 minutes the method allows rapid detection of Dicyandiamide.| Market: | Pharma |
| Keywords: | HPLC, Hypersil GOLD aQ, UV detection, Dicyandiamide |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific, APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.