Find methods for your needs
AN1157: Determination of Organic Acids in Kombucha Using a High-Pressure Ion Chromatography System
Description
This work used the properties of the 4-μm-particle-size Dionex IonPac AS11-HC-4μm column, combined with the Thermo Scientific™ Dionex™ ICS-5000+ HPIC system, to produce a high-resolution separation of organic acids and inorganic anions in kombucha samples, enabling accurate determinations of those analytes. An ICS-6000 can be used for this application.| Market: | Food and Beverage |
| Keywords: | Dionex ICS-5000+, HPIC, Suppressed Conductivity, Dionex IonPac AS11-HC-4μm column, ICS-6000 |
| Matrix: | Fermented beverage |
| Author: | Beibei Huang; Jingli Hu; Jeffrey Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.