Find methods for your needs
Rapid LC-MS/MS method for the determination of enalapril and enalaprilat from human plasma using Thermo Scientific SOLA SPE and a Thermo Scientific Hypersil GOLD HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of enalapril and enalaprilat. The sample is prepared using Thermo Scientific SOLA solid phase extraction (SPE) cartridges, and the separation is performed on a Thermo Scientific Hypersil GOLD HPLC column, with MS/MS detection. For Research Use Only. Not for use in diagnostic procedures.
Market: Pharma
Keywords: Benazepril, Enalapril, Enalaprilat, Human Plasma, Hypersil GOLD, LC-MS/MS, MS detection, SOLA, SPE
Matrix: Human plasma
Uploaded on 8/22/2012.
For Research Use Only. Not for use in diagnostic procedures.