Find methods for your needs
Rapid analysis of estradiol and estrone using a Thermo Scientific Accucore RP-MS HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of estradiol and estrone. The separation is performed on a Thermo Scientific Accucore RP-MS HPLC column in less than 1 minute, with UV detection at 220 nm.Market: | Pharma |
Keywords: | Accucore RP-MS, Estradiol, Estrogens, Estrone, Hormones, HPLC, UV detection |
Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.