Find methods for your needs
Assay and organic impurities analysis of Cefotaxime sodium using a C18 HPLC column using USP 39 monograph - (procedure 1)
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the assay and organic impurities (procedure 1) analysis of Cefotaxime Sodium. The analysis was performed on a Thermo Scientific Syncronis aQ HPLC column using the method described in the USP 39. The results obtained for resolution and tailing factor met the criteria stated in the USP. The results obtained for resolution between the peaks due to cefetamet and Cefotaxime related compound E was 8.9 (USP criteria of minimum 1.5) and tailing factor for cefotaxime was 1.4 (USP criteria of maximum 2).| Market: | Pharma |
| Keywords: | Cefotaxime, HPLC, Syncronis aQ, UV detection, USP 39 monograph, beta-lactam antibiotic |
| Matrix: | N/A |
| Author: | Krishnarao Dara |
| Affiliation: | APAC CoE Lab, Ahmedabad, India |
For Research Use Only. Not for use in diagnostic procedures.