Find methods for your needs
AN1100: Determination of Benzo[a]pyrene in Municipal Drinking Water Using Automated Solid-Phase Extraction and Liquid Chromatography with Fluorescence Detection
Description
A Dionex AutoTrace 280 solid-phase extraction instrument was used to reduce analyst intervention and automatically complete SPE column activation, sample loading, and the elution and collection of Benzo[a]pyrene. The eluted samples were solvent exchanged and concentrated to a defined volume, and then trace Benzo[a]pyrene in municipal water was measured using HPLC with fluorescence detection.| Market: | Environmental |
| Keywords: | Polyaromatic hydrocarbons, Acclaim C18, Dionex AutoTrace 280, HPLC-FLD, GB5749-2006, HJ 478-2009 |
| Matrix: | Drinking water |
| Author: | Hongguo Zheng, Mingle Ye, Lina Liang |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.