Find methods for your needs
Fast analysis of 14 positional isomers using a Thermo Scientific Accucore PFP HPLC column Be the first to rate this application
Description
The Thermo Scientific HPLC system is applied for the analysis of 14 positional isomers. The separation is performed on a Thermo Scientific Accucore PFP HPLC column, with UV detection at 270 nm.
Market: Chemical
Keywords: Accucore PFP, HPLC, Positional isomers, UV detection
Matrix: N/A
Uploaded on 8/22/2012.
For Research Use Only. Not for use in diagnostic procedures.