Find methods for your needs
Rapid determination of lamivudine using the USP method and a Thermo Scientific Accucore aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the analysis of lamivudine. The separation is performed on a Thermo Scientific Accucore aQ HPLC column in less than 5 minutes, with UV detection at 277 nm. The method of analysis was adapted from the USP monograph, by using an in-house method transfer calculator.| Market: | Pharma |
| Keywords: | Accucore aQ, HPLC, Lamivudine, Method transfer, USP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.