Find methods for your needs
USP 39 monograph - Organic impurities analysis of Cefdinir using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the organic impurities analysis of cefdinir. The analysis was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the USP 39 monograph. The results obtained for resolution and %RSD met the criteria stated in the USP 39. The resolution between cefdinir and the third peak of cefdinir related compound A was 3.1 (USP criteria of NLT 1.5) and %RSD for cefdinir was 0.1% (USP criteria of NMT 2.0%).| Market: | Pharma |
| Keywords: | Acclaim 120 C18, Cephalosporin Antibiotic, HPLC, UV detection, Cefdinir, USP 39 monograph |
| Matrix: | N/A |
| Author: | Krishnarao Dara |
For Research Use Only. Not for use in diagnostic procedures.