Find methods for your needs
Rapid analysis of zidovudine using a Thermo Scientific Accucore C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of Zidovudine. The separation is performed on a Thermo Scientific Accucore C18 HPLC column in under 3 minutes, with UV detection at 265 nm.| Market: | Pharma |
| Keywords: | Accucore C18, Antiretroviral, Azidothymidine, HPLC, Retrovir, UV detection, Zidovudine |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.