Find methods for your needs
Determination of lamivudine using the USP method and a Thermo Scientific Syncronis aQ HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of lamivudine using the USP method. The separation is performed on a Thermo Scientific Syncronis aQ HPLC column, with UV detection at 277 nm.| Market: | Pharma |
| Keywords: | HPLC, Lamivudine, nRTI, Nucleoside analog reverse transcriptase inhibitor, Syncronis aQ, USP, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.