Find methods for your needs
AN335: Accelerated Solvent Extraction (ASE) of Active Ingredients from Natural Products
Description
Herbal products, nutraceuticals, and other food supplements are subject to increasingly rigorous investigation and development. Industry standardization, as well as labeling requirements under new federal regulations, have prompted tighter regulation and monitoring of the active ingredients in these products . This Application Note describes the use of ASE for extraction of commercially available nutritional supplements, specifically hypericin from Hypericum perforatum (St. John's wort) and berberine from Hydrastis canadenis (goldenseal root).| Market: | Food and Beverage; Pharma |
| Keywords: | Berberine, ASE, ASE 200, ASE 350, ASE Prep DE, Soxhlet Extraction, Herbal products, hypericin |
| Matrix: | Nutritional supplements |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.