Find methods for your needs
Improved analysis of triazines using a Thermo Scientific Syncronis C18 HPLC column
Description
The Thermo Scientific HPLC system is applied for the determination of triazines. The separation is performed on a Thermo Scientific Syncronis C18 HPLC column, with UV detection at 280 nm.| Market: | Environmental |
| Keywords: | Ametryn, Atrazine, HPLC, Prometryn, Propazine, Simazine, Simetryn, Syncronis C18, Triazines, UV detection |
| Matrix: | N/A |
For Research Use Only. Not for use in diagnostic procedures.