Find methods for your needs
Measurement of Vitamin D in Milk and Infant Formulas Using Automated Online Sample Preparation with Liquid Chromatography/ Tandem Mass Spectrometry
Description
Vitamin D, including vitamin D2 and D3 (Figure 1), is essential to calcium absorption and bone health. Current studies suggest vitamin D could help prevent a wide range of disorders. Although vitamin D can be synthesized by the human body from exposure to sunlight, fortification in some dietary sources becomes necessary. We describe an easy and comprehensive LC-MS/MS method using a Transcend TLX-1 system powered by Thermo Scientific TurboFlow technology to measure vitamin D levels in infant formula. TurboFlow technology eliminates most traditional sample preparation steps.Market: | Food and Beverage |
Keywords: | Milk, Infant Formula, TurboFlow Technology, Vitamin D, Transcend TLX-1, TSQ Access MAX |
Matrix: | Infant formula |
Author: | Yang Shi, Catherine Lafontaine, Matthew Berube, John Fink, François Espourteille |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.