Find methods for your needs
Analysis of Melamine and Cyanuric Acid in Food Matrices by LC-MS/MS
Description
In March 2007, several North American manufacturers of pet food voluntarily issued nationwide recall notices for some of their products that were reportedly associated with renal failure in pets. It was discovered that melamine-contaminated China-sourced ingredients had been used to prepare feed for chickens, swine, and catfish. Consequently, the FDA and the USDA have developed methods for the analysis of melamine residues in animal tissue. Both methods use tandem MS detection and employ disposable strong cation exchange SPE cartridges to prepare samples for liquid chromatographic analysis.| Market: | Food and Beverage |
| Keywords: | LC-MS/MS, Cyanuric Acid, Food Safety, TSQ Quantum Ultra |
| Matrix: | Feed |
| Author: | PeterVarelis, Jonathan Beck, Kefei Wang, and Dipankar Ghosh |
| Affiliation: | National Center for Food Safety and Technology, Illinois Institute of Technology; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.