Find methods for your needs
Increasing Speed of UHPLC-MS Analysis Using Single-stage Orbitrap Mass Spectrometer
Description
Productivity of an LC-MS system is measured in samples per day. Modern ultrahigh pressure LC-MS (UHPLC-MS) methods increasingly deal with very short gradients, leading to chromatographic signals with peak widths below 5 seconds at the base. It is still a challenge for high-resolution, accurate mass (HR/AM) systems to provide a sufficient number of scans (=10) across the chromatographic peak in full scan mode without compromising sensitivity and selectivity. We describe a system that addresses these challenges.| Market: | Environmental |
| Keywords: | HRAM, Pesticide Screening, Exactive Plus, Feed analysis |
| Matrix: | Animal feed |
| Author: | Olaf Scheibner and Maciej Bromirski |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.