Find methods for your needs
Fast and Ultrafast LC-MS/MS Methods for Robust and Reliable Analysis of Pesticides in Food Using the Vanquish UHPLC System
Description
Food safety is an increasing concern that has resulted in stringent pesticide regulation globally and in continuous recalls of food products. Food safety regulations require the screening and the quantitation of a large number of pesticides in food at maximum residue levels generally set in the ppb-ppm range to minimize their possible negative effects on human health. Here we present a comparison between fast and ultrafast LC-MS/MS methods in timed SRM mode for the analysis of 250+ pesticides in food extracts. The two methods were compared in terms of analysis time and data quality.| Market: | Food and Beverage |
| Keywords: | Food Safety, Vanquish UHPLC System, Retention Time Reproducibility, Timed SRM, High-throughput |
| Matrix: | Foods |
| Author: | Giorgia Greco, Claudia P.B. Martins, Katerina Bousova, and Remco Swart |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.