Find methods for your needs
Multi-mycotoxin Screening and Quantitation Using UHPLC, High Resolution and Accurate Mass
Description
Mycotoxins are the toxic secondary metabolites produced by many species of microscopic filamentary fungi occurring on field cereals, including barley. There is a trend toward the simplification of sample preparation procedures as much as possible. Full spectral data acquisition techniques are also preferred because of their ease of usage, along with the possibility of retrospective archived data mining. We introduce a multi-mycotoxin method for analysis of 32 mycotoxins in beer based on very simple sample preparation and UHPLC coupled with full spectral Orbitrap MS detection.| Market: | Food and Beverage |
| Keywords: | Beer, Accela UHPLC, cereals, Exactive LC-MS |
| Matrix: | Beer |
| Author: | Milena Zachariášová, Jana Hajšlová, Michal Godula |
| Affiliation: | Institute of Chemical Technology - Czech Republic; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.