Find methods for your needs
Highly Selective Detection and Identification of Nitrofuran Metabolites in Honey Using LC-MS/MS
Description
Nitrofurans are broad-spectrum antibiotics used to treat bees and other animals with bacterial infections. Nitrofurans have been banned from use in food-producing animals in several countries. Several studies have shown that animals rapidly metabolize nitrofurans within a few hours so detection has focused on the metabolites rather than the native drug. We describe a method to accurately quantitate nitrofuran metabolites at levels as low as 0.3 ppb in a matrix consisting of honey using the TSQ Quantum Discovery.| Market: | Food and Beverage |
| Keywords: | LC-MS/MS, Surveyor HPLC, Quantitative Analysis, TSQ Quantum Discovery |
| Matrix: | Honey |
| Author: | Eduardo Matus, Jean-Jacques Dunyach, Alejandro Albornoz |
| Affiliation: | Food Science Laboratories - Argentina; Eidomet - Argentina; Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.