Find methods for your needs
Identification of Psychotropic Substances in Mushrooms and Chocolate by UHPLC/MS Be the first to rate this application
identification psychotropic substances mushrooms chocolate by uhplcms
# Compound Name Compound Class RT (min)
1 Psilocybin Psychotropic 0.65
2 psilocin Psychotropic 2.25
Description
The identification of controlled substances in food matrices is a challenge for forensic laboratories. Classical techniques, such as color tests and thin-layer chromatography, do not provide molecular structural information and cannot be used as a principal means of identification. Infrared spectroscopy (IR) and gas chromatography/mass spectrometry (GC/MS) can identify controlled substances, but suffer from several shortcomings. We describe a UHPLC/MS method for the detection, separation and confirmation of psilocybin and psilocin in mushrooms and chocolate matrices.
Market: Pharma
Keywords: Toxicology, Forensics, Magic Mushroom, Accela UHPLC System, Psilocybin, Psilocin
Matrix: Chocolate, Mushrooms
Author: Jason R. Stenzel, Guifeng Jiang, Chris Loran
Affiliation: Washington State Patrol – Crime Laboratory Division-USA, Thermo Fisher Scientific
Uploaded on 3/6/2016.
For Research Use Only. Not for use in diagnostic procedures.