Find methods for your needs
Determination of Bisphenol A in Infant Formula by Automated Sample Preparation and Liquid Chromatography-Mass Spectrometry
Description
2,2-bis(4-hydroxyphenyl) propane, commonly known as Bisphenol A (BPA), is one of the primary chemicals used to make plastics. It is also heavily used in the production of various types of food and drink containers. Because BPA has been known to leach from the plastic lining of metal canned food, the potential risks of exposure to BPA have been a great concern over the past few years. We describe a six-minute LC-MS/MS method using automated sample preparation for the assay of BPA in canned infant formula powder by negative ion APCI using a deuterated internal standard (BPA-d16).| Market: | Food and Beverage |
| Keywords: | Food Safety, Infant Formula, TurboFlow Technology, Aria TLX-1 |
| Matrix: | Infant formula |
| Author: | Yang Shi, Catherine Lafontaine, Matthew Berube, Francois Espourteille |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.