Find methods for your needs
Analysis of Chloroacetanilide and Other Acetamide Herbicide Degradates in Drinking Water by LC-MS/MS
Description
Chloroacetanilide herbicide compounds in drinking water, specifically ethanesulfonic acid (ESA) and oxanillic acid (OA) degradates, may be found in water supplies near agricultural production areas and are used on various crops including corn and soybeans. Therefore, monitoring of these compounds is of interest to various regulatory agencies around the world. This application note illustrates the direct analysis of drinking water spiked with chloroacetanilide herbicide compounds. Excellent linearity and reproducibility are demonstrated over the entire calibration range.| Market: | Environmental |
| Keywords: | Herbicides, Pesticides |
| Matrix: | Drinking water |
| Author: | Jonathan Beck |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.