Find methods for your needs
Fast and Accurate Determination of Algal Toxins in Water Using Online Preconcentration and UHPLC-Orbitrap Mass Spectrometry
Description
When the density of the colonies of Microcystis and Nodularia cyanobacteria surpass a certain level, they produce hepatotoxic substances called microcystins and nodularins, respectively,2 while Anabaena and Apha-zinomenon are known to produce a neurotoxin called anatoxin. We describe a column-switching technique based on online preconcentration and high-resolution, full-scan Thermo Scientific™ Orbitrap™ mass spectrometry to obtain fast and accurate results for the determination of algal toxins in drinking water.| Market: | Environmental |
| Keywords: | Water analysis, Exactive, Orbitrap, EQuan, Algal toxins, Cyanobacteria, Online concentration |
| Matrix: | Water |
| Author: | Jaewon Choi, Je-Heon Jang, Jennifer Massi, Jonathan Beck |
| Affiliation: | Thermo Fisher Scientific; Water Analysis and Research Center-Korea |
For Research Use Only. Not for use in diagnostic procedures.