Find methods for your needs
Fast and Accurate Identification of Pesticides by Direct Analysis in Real Time (DART) Ionization with Orbitrap Mass Spectrometry
Description
We describe a method incorporating direct analysis in real time (DART) ionization and Thermo Scientific Orbitrap high-resolution mass spectrometry for rapid analysis and identification of contaminating substances in water.| Market: | Environmental |
| Keywords: | Pesticides, Water analysis, Exactive, Direct Analysis in Real Time ionization (DART), Orbitrap |
| Matrix: | Water |
| Author: | Jaewon Choi, Wonseok Choi, Jennifer Massi, Mari Prieto Conaway |
| Affiliation: | Thermo Fisher Scientific; Water Analysis & Research Center - Korea |
For Research Use Only. Not for use in diagnostic procedures.