Find methods for your needs
Targeted Screening of Fungal and Plant Metabolites in Wheat, Corn, and Animal Feed Using Automated Online Sample Preparation Coupled to Orbitrap LC-MS
Description
We describe a method that can be applied, for targeted screening of 21 fungal and plant metabolites with automated online sample cleanup utilizing a Thermo Scientific Transcend system coupled to a Thermo Scientific Exactive high-resolution mass spectrometer. This method has been validated according to current EU Commission legislation (2006, 2007).| Market: | Food and Beverage |
| Keywords: | Food, TurboFlow Technology, Transcend system, Exactive HRMS, Fungal Metabolites, Plant Metabolites |
| Matrix: | Grain |
| Author: | Ebru Ates, Michal Godula, and Klaus Mittendorf |
| Affiliation: | Food Safety Response Center, Dreieich, Germany |
For Research Use Only. Not for use in diagnostic procedures.