Find methods for your needs
Detection of Glucocorticoid Residues in Animal-derived Food by HPLC-MS/MS Be the first to rate this application
detection glucocorticoid residues animalderived food by hplcmsms
# Compound Name Compound Class RT (min)
1 Prednisolone Glucocorticoids 6.89
2 Hydrocortisone Glucocorticoids 7.20
3 Prednisone Glucocorticoids 7.22
4 Methylprednisolone Glucocorticoids 12.45
5 Dexamethasone Glucocorticoids 13.41
6 Betamethasone Glucocorticoids 14.44
7 Beclomethasone Glucocorticoids 17.95
8 Fludrocortisone acetate Glucocorticoids 25.33
Description
We developed an effective LC-MS/MS method to detect eight gluococorticoid residues in the liver and muscles of pig, cattle, and lamb, as well as in chicken, eggs and milk.
Market: Food and Beverage
Keywords: TSQ Quantum, Veterinary drugs, Food residue analysis, SRM (Selective Reaction Monitoring)
Matrix: Livestock
Author: Chen Dongmei, Fei Liu, and Charles Yang
Affiliation: Thermo Fisher Scientific; National Reference Laboratory of Veterinary Drug Residues - China
Uploaded on 2/24/2016.
For Research Use Only. Not for use in diagnostic procedures.