Find methods for your needs
Sensitive Determination of Hexavalent Chromium in Drinking Water Using a Compact Ion Chromatography System
Description
This record demonstrates that the method in AU179 can be run successfully using an Integrion Ion Chromatography (IC) system. AU179 describes modification of the conditions described in EPA Method 218.6, including use of the column in the 2 mm format and a smaller reaction coil to increase method sensitivity. The modified method, which is now US EPA Method 218.7, uses a Dionex IonPac AS7 column (2 × 250 mm), a 1000 μL injection volume, and postcolumn reaction (using a 125 μL reaction coil) followed by visible absorbance detection at 530 nm.| Market: | Environmental |
| Keywords: | hexavalent chromium, EPA Method 218.6, EPA Method 218.7, Drinking water, Integrion, UV Absorbance Detection |
| Matrix: | Drinking water |
| Author: | Jingli Hu; Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific, Sunnyvale, CA USA |
For Research Use Only. Not for use in diagnostic procedures.