Find methods for your needs
食醋中安赛蜜的检测 Be the first to rate this application
食醋中安赛蜜的检测
Description
本方法通过优选色谱柱,最终确定使用WAX-1 色谱柱,以阴离子交换结合反相分离的混合模式,以磷酸盐- 乙腈洗 脱系统实现了安赛蜜色谱峰与基质成份的良好分离,无需 复杂前处理过程(简单稀释过滤即可),方法快速简便(测 试周期约为10min),灵敏度高(检测限按供试品取样量 折算可达0.8 μg/ml,而GB/T 5009.140-2003 的方法检测限为 4 μg/ml),精密度和回收率好,为食醋类样品中安赛蜜的 测定提供了较好的方法参考。
Market: Food and Beverage
Keywords: 高效液相色谱;乙酰磺胺酸钾;WAX-1 色谱柱
Matrix: 食醋
Author: 冉良骥
Uploaded on 1/31/2016.
For Research Use Only. Not for use in diagnostic procedures.