Find methods for your needs
Purity of the Liposomal Biotherapeutic Delivery Compound, DC-Cholesterol, by HPLC-CAD
Description
Cationic Lipids are used as a transfection agent in biotechnological products created in the siRNA market space. As part of the lipid-based delivery system for these bio-therapeutic agents, their quality needs to be determined. DC-Cholesterol, 3ß-[N-(Dimethylaminoethane)carbamoyl]cholesterol, is such a lipid, and using the Vanquish UHPLC with the universal Corona charged aerosol detector, many impurities were separated and can be quantified.<enter final 90 characters of description>| Market: | Other |
| Keywords: | Cationic lipids, DC-cholesterol, siRNA |
| Matrix: | Methanol / THF (1:1) |
For Research Use Only. Not for use in diagnostic procedures.