Find methods for your needs
Determining Nitrosamines Using GC-MS/MS with Electron Ionization
                                Description
The highly sensitive TSQ Duo triple quadrupole mass spectrometer can analyze nitrosamines in EI mode. Using automatic selected reaction monitoring (AutoSRM) software to fully optimize the choice of transitions and collision energies, equivalent detection limits by EPA method can be achieved without using a large volume injection.| Market: | Environmental | 
| Keywords: | Nitrosamines, GC-MS/MS, AutoSRM, Electron Energy, Triple Quadrupole | 
| Author: | David Steiniger and Alexander Semyonov | 
| Affiliation: | Thermo Fisher Scientific | 
For Research Use Only. Not for use in diagnostic procedures.