Find methods for your needs
Fast Determination of Anions in Bottled Water Be the first to rate this application
Description
Inorganic anions in mg/L concentrations in bottled water sample were seperated using a 2 x 150 mm, IonPac AS18-Fast anion-exchange column designed for fast separation using electrolyticallly generated hydroxide eluents.
Market: Environmental
Matrix: Bottled water
Uploaded on 7/8/2011.
For Research Use Only. Not for use in diagnostic procedures.