Find methods for your needs
Determination of Anions in an Artificial Volcanic Gas Condensate Sample on 2 mm IonPac AS25 Column
Description
Determination of Anions in an Artificial Volcanic Gas Condensate Sample on 2 mm IonPac AS25 Column. This sample simulates a volcanic gas condensate sample containing 100 mg/L carbonate, 10 mg/L sulfate and ppb to ppm concentrations of halides, sulfite, thiocyanate, and thiosulfate.| Market: | Environmental |
| Matrix: | Volcanic gas condensate |
For Research Use Only. Not for use in diagnostic procedures.