Find methods for your needs
Analysis of Alkylphenols Using GC-MS/MS and Automated SRM Development
Description
This application describes the analysis of alkylphenol compounds in food and environmental matrices, illustrating the productivity and high-quality results from the Thermo Scientific TSQ DUO Triple Quadrupole MS system with automated selected reaction monitoring (SRM) development. AutoSRM was used as useful tool for automated MRM method development and maintenance, delivering an optimized method without tedious and laborious procedures. The Chromeleon CDS software offers excellent processing options for data reviewing, including ion ratio checks and reporting.| Market: | Environmental; Water Analysis; Food and Beverage |
| Keywords: | AutoSRM, Chromeleon Chromatography Data System, Alkylphenols, Isomer integration, Method Development, Nonylphenol, NP, NPEs, Water Framework Directive |
| Matrix: | Food, Water |
| Author: | Inge de Dobbeleer, Joachim Gummersbach, Hans-Joachim Huebschmann |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.