Find methods for your needs
A Fast and Accurate Method to Monitor Protein PEGylation Reactions by HPLC-UV-CAD
Description
This application provides a fast and accurate way for biopharmaceutical chemists to monitor PEGylation reactions by using HPLC with complementary UV/Vis and charged aerosol detection. A high resolution separation on the MAbPac RP resolves PEGylation reagents, unreacted insulin, and PEGylated insulin products that differ in the number and sites of attached PEG.| Market: | Other |
| Matrix: | Reaction mixture |
| Author: | Dave Thomas |
| Status: | Proof of Concept |
For Research Use Only. Not for use in diagnostic procedures.