Find methods for your needs
TN23: Ion Chromatography of Lanthanide Metals (Part 3/3: Simultaneous Separation of Transition Metals and Lanthanide Metals)
Description
A class separation of lanthanide metals from transition metals is achieved by taking advantage of the electroselectivity differences between transition metal complexes and lanthanide metal complexes. Transition metals form stable monovalent or divalent anionic complexes with PDCA, while lanthanide elements form stable trivalent anionic complexes with PDCA.| Market: | Chemical |
| Keywords: | Ion Chromatography, Lanthanide Metals, Transition Metals |
| Matrix: | Water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.