Find methods for your needs
Anions and Organic Acids in Young Coconut water by High Pressure Capillary IC on a monolith capillary column Be the first to rate this application
Description
Anions and organic acids in young coconut water by High Pressure Capillary IC on IonSwift MAX-100 column. This application uses the advantages of speed of monolith column technology while enjoying the chemistry of IonPac AS11 which is optimized for organic acid separations. While it is not necessary, this application is run on a high pressure capillary ICS-5000 IC system.
Market: Food and Beverage
Keywords: Chloride, Malate, Quinate, Citrate, Aconitate, IonSwift MAX-100 column, anions, Phosphate, Isocitrate, RFIC, Sulfate, monolith, capillary IC
Matrix: Coconut water
Uploaded on 5/8/2011.
For Research Use Only. Not for use in diagnostic procedures.