Find methods for your needs
TN10: Determination of Transition Metals by Ion Chromatography Using Oxalate Buffer Eluent
Description
The IonPac CS5A column has both cation and anion exchange capacity, allowing metals to be separated as cations or anions on a single column. This is called a mixed mode separation.Transition metals are separated as both cationic and anionic complexes with the oxalate chelating agent added to the eluent.The metals are detected by measuring the absorbance at 530 nm of the complex formed with the postcolumn PAR reagent.| Market: | Chemical |
| Keywords: | Transition Metals, Ion Chromatography, IonPac CS5A Column, Oxalate Chelating Agent, Postcolumn PAR reagent |
| Matrix: | Water |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.