Find methods for your needs
Impurity analysis of loratadine formulation using HPLC-UV with an ion-pair method
Description
The Thermo Scientific Dionex Ultimate 3000 HPLC system was applied for the impurity analysis of loratadine in formulation. The Thermo Scientific Hypersil GOLD HPLC column performed the separation of loratadine and its impurities in 55 minutes with UV detection at 254 nm.| Market: | Pharma |
| Keywords: | HPLC, Hypersil GOLD, UV detection, Loratadine |
| Matrix: | Formulation |
| Author: | Prakash Chander |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.