Find methods for your needs
EP 8.0 monograph - impurity analysis of mefenamic acid using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of mefenamic acid and its impurities. The separation was performed on a Thermo Scientific Hypersil GOLD column using the method described in the EP 8.0 monograph. Resolution between impurity C and impurity D in reference solution (b) was 4.5 (exceeding EP criteria of minimum 3.0). Retention time of mefenamic Acid was 8.16 minutes (meeting EP criteria of about 8 minutes). Relative retention time criteria was also exceeded.| Market: | Pharma |
| Keywords: | Hypersil GOLD, Mefenamic acid, UV detection, EP 8.0 monograph |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.