Find methods for your needs
USP 38 monograph - assay analysis of mefenamic acid using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the assay analysis of mefenamic acid. The analysis was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the USP 38 monograph. The results for column efficiency for the standard solution was 15158 (exceeding the USP criteria of not less than 8200). The tailing factor results for mefenamic acid in standard solution was 0.83 (USP criteria not more than 1.6). The % RSD for replicate injection of standard solution was 0.19% (exceeding the USP criteria not more than 1.0%)| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, Mefenamic acid, UV detection, USP 38 monograph |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.