Find methods for your needs
AU162: Determination of Biogenic Amines in Fruit, Vegetables, and Chocolate Using Ion Chromatography with Suppressed Conductivity and Integrated Pulsed Amperometric Detections
Description
In this update, the procedures described in AN 183 are used to determine biogenic amine levels in kiwi fruit, spinach, and chocolate. Application Note 183 (AN 183) describes the use of the IonPac CS18, a weak acid cation-exchange column, with suppressed conductivity, integrated pulsed amperometric detection (IPAD), and UV for the detection of underivatized biogenic amines in meat and cheese. The CS18 allows separation of biogenic amines without the use of highly-concentrated acidic eluents or organic solvents while still providing resolution of closely eluting peaks.| Market: | Food and Beverage |
| Keywords: | IPAD, RFIC, AN 183, Histamine, Tyramine, AU162, IonPac CS18, Putrescine, Cadaverine |
| Matrix: | Food extract |
| Author: | Brian De Borba and Jeff Rohrer |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.