Find methods for your needs
EP 8.0 monograph - assay analysis of cefalexin using reversed-phase HPLC-UV
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the assay analysis of cefalexin. The separation was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the EP 8.0 monograph. The results obtained for resolution exceeded the criteria stated in the EP. Resolution between cefalexin and cefradine was 14.1 (EP criteria of minimum of 4.0).| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, UV detection, EP 8.0 monograph, Cefradine, Cefalexin |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.