Description
The Thermo Scientific Trace 1310 ISQ system is applied for the direct injection of extracted organophosphorus pesticide samples. Separation is performed on a Thermo Scientific TraceGOLD TG-5SilMS column with MS SIM detection.Market: | Environmental |
Keywords: | GC-MS, Pesticides, Organophosphorus pesticides, MS detection, OPP |
Matrix: | N/A |
Author: | Richard Law |
Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.