Find methods for your needs
USP 38 monograph - impurities test for levofloxacin using a C18 HPLC column
Description
The Thermo Scientific Dionex UltiMate 3000 LC system is applied for the analysis of levofloxacin and its impurities. The separation was performed on a Thermo Scientific Acclaim 120 C18 HPLC column using the method described in the USP 38 monograph. The results obtained for % relative standard deviation and signal to noise ratio exceeded the criteria stated in the USP. Relative standard deviation for system suitability solution was 0.1% (USP criteria of not more than 1.0%) and signal to noise ratio for sensitivity solution was 58 (USP criteria not less than 10).| Market: | Pharma |
| Keywords: | Acclaim 120 C18, HPLC, UV detection, USP 38 monograph, Levofloxacin |
| Matrix: | N/A |
| Author: | Manish Singh |
| Affiliation: | Thermo Fisher Scientific |
For Research Use Only. Not for use in diagnostic procedures.